Learn More
Cayman Chemical Nitro Blue Tetrazolium chlor
SDP
Catalog No. 502968192
Encompass_Preferred
A chromogenic substrate that, like other tetrazolium compounds, can be reduced to produce a colored formazan derivative; used in the “NBT test” to evaluate the activity of NADPH oxidase in phagocytes; paired with PCIB for the colorimetric detection of alkaline phosphatase activity
Catalog No. 50-296-8192 Supplier Cayman Chemical Supplier No. 17341250 MG
A chromogenic substrate that, like other tetrazolium compounds, can be reduced to produce a colored formazan derivative; used in the “NBT test” to evaluate the activity of NADPH oxidase in phagocytes; paired with PCIB for the colorimetric detection of alkaline phosphatase activity