Learn More
Electron Microscopy Sciences Nitro Blue Tetrazolium Chloride 100 MG
SDP
Catalog No. 5027498
Encompass_Preferred
(Nitro, BT, p-Nitrotetrazolium Blue; 2, 2´-Di-p-nitrophenyl-5, 5´diphenyl-[3, 3´-dimethoxy-4, 4´-diphenylene] ditetrazolium chloride)
CAS #298-83-9 C₄₀H₃₀Cl₂N₁₀O₆ F.W. 817.65 g/mol
A substrate for dehydrogenases and other oxidases. Also available in tablet form. Tablet contains 10mg substrate per tablet.
Catalog No. 50-274-98 Supplier Electron Microscopy Sciences Supplier No. 1903501
(Nitro, BT, p-Nitrotetrazolium Blue; 2, 2´-Di-p-nitrophenyl-5, 5´diphenyl-[3, 3´-dimethoxy-4, 4´-diphenylene] ditetrazolium chloride)
CAS #298-83-9 C₄₀H₃₀Cl₂N₁₀O₆ F.W. 817.65 g/mol
A substrate for dehydrogenases and other oxidases. Also available in tablet form. Tablet contains 10mg substrate per tablet.