Learn More
Crescent Chemical Co Inc NITRO BLUE TETRAZOLIUM CHLORID
SDP
Supplier:  Crescent Chemical Co Inc SE30550.03
Encompass_Preferred
Nitro Blue tetrazolium chloride (NBT) analytical grade A ditetrazolium compound for demonstrating succinic dehydrogenase activity (1), glucose-6-phosphate dehydrogenase (2) and xanthine oxidase (3). Used in conjuction with BCIP (cat. no. 15247) for detection of alkaline phosphatase.Sto
Catalog No. 50-247-936