Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride monohydrate, 90%

Catalog No. AC128590050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC128590050 5 g
AC128590010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128590050 Supplier Thermo Scientific Chemicals Supplier No. 128590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.644 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitro Blue Tetrazolium Chloride monohydrate
CAS 298-83-9, 67-56-1, 7732-18-5
CAS Min % 88.0
CAS Max % 100.0
Infrared Spectrum Authentic
Loss on Drying 10% max. (105°C)
Packaging Glass bottle
Assay Percent Range 90%
Molecular Formula C40H30Cl2N10O6
MDL Number MFCD00150013
Color Yellow
Synonym Nitro BT, Nitro blue tetrazolium
Quantity 5 g
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Physical Form Powder
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
Molecular Weight (g/mol) 817.644
PubChem CID 9281
ChEBI CHEBI:9505
Formula Weight 867.69
Percent Purity ≥88%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.