Learn More
Research Products International Corp NBT [Nitro Blue Tetrazolium] [2,2'-Di-P-Nitrophenyl-5, 5'-Diphenyl-3, 3'-(3,3'-Dimethoxy-4, 4'-Diphenylene)-Ditetrazolium Chloride], 1 Gram
SDP
Catalog No. 50213569
Encompass_Preferred
Colorimetric substrate for dehydrogenases and other oxidases. NBT is used with X-Phos for colorimetric detection of alkaline phosphatase activity in blotting, immunohistochemical and cytochemistry techniques. Protect from light.
Catalog No. 50-213-569 Supplier Research Products International Corp Supplier No. N660001.0
Colorimetric substrate for dehydrogenases and other oxidases. NBT is used with X-Phos for colorimetric detection of alkaline phosphatase activity in blotting, immunohistochemical and cytochemistry techniques. Protect from light.