Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitro Blue Tetrazolium, Sigma-Aldrich

Catalog No. 01259043 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 Tablets
25 Tablets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-043 25 Tablets
01-259-044 10 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-043 Supplier MilliporeSigma Supelco Supplier No. N551425TAB
Add to Cart
Add to Cart

Nitro Blue Tetrazolium is a tetrazole compound with two tetrazole rings.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitro Blue Tetrazolium
CAS 298-83-9
Color Yellow
pH 4.5 to 5.5
Quantity 25 Tablets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.