Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nitro-Grela 97.0+%, TCI America™
SDP

Catalog No. N1060500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1060500MG 500 mg
N1060100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1060500MG Supplier TCI America Supplier No. N1060500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 502964-52-5
Molecular Formula C31H37Cl2N3O3Ru
MDL Number MFCD28411645
Synonym [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro[(2-isopropoxy-5-nitrobenzylidene)]ruthenium(VI)

Specifications

Quantity 500 mg
Formula Weight 671.63
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material nitro-Grela
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.