Learn More
Dojindo Molecular Technologies Inc NITRO-TB (1G), CAS: 298-83-9, Dehydrogenase detection, staining, used for dehydrogenase detection on agarose gels.
SDP
Catalog No. 501903740
Encompass_Preferred
Nitro-TB is dissolved by hot water or hot methanol. Other organic solvents such as acetone and ether cannot solubilize Nitro-TB. Nitro-TB is easily reduced by dehydrogenase to purple formazan dye aggregates (λmax=530 nm, ε=3.6 x 104). It is used for dehydrogenase detection on agarose gels.
Catalog No. 50-190-3740 Supplier Dojindo Molecular Technologies Inc Supplier No. N01112
Nitro-TB is dissolved by hot water or hot methanol. Other organic solvents such as acetone and ether cannot solubilize Nitro-TB. Nitro-TB is easily reduced by dehydrogenase to purple formazan dye aggregates (λmax=530 nm, ε=3.6 x 104). It is used for dehydrogenase detection on agarose gels.