Learn More
American Radiolabeled Chemicals Inc NITROACEPHE 14C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 359750UC
Encompass_Preferred
p-Nitroacetophenone-carbonyl-14C 50uCi in ethanol, 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 165.15, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-963