Learn More
Abcam NBD-dodecanoic acid, Fatty acid ligand binding site probe, 100MG
Supplier:  Abcam AB145361100MG
Encompass_Preferred
MW 378.42 g/mol, Purity >98%. Suitable for probing the ligand binding sites of fatty acid and sterol carrier proteins as well as the investigation of lipolysis processes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-207