Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene-d5, for NMR, 99 atom % D

Catalog No. AC166360050
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166360050 5 g Glass Bottle
AC166360100 10 g Glass Bottle
AC166360250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166360050 Supplier Thermo Scientific Chemicals Supplier No. 166360050
Add to Cart

CAS: 4165-60-0 | C6H5NO2 | 128.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-60-0
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 128.14
MDL Number MFCD00044415
InChI Key LQNUZADURLCDLV-RALIUCGRSA-N
Synonym nitrobenzene-d5, benzene-d5, nitro, nitrobenzene d5, benzene-d5-, nitro, 1-nitro 2 h? benzene, nitrobenzene-d5, 99.5 atom % d, nitro 2h5 benzene, 1-nitro 2h5 benzene, 2h5 benzene, nitro, nitrobenzene-d5, analytical standard
PubChem CID 123210
IUPAC Name 1,2,3,4,5-pentadeuterio-6-nitrobenzene
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])[N+]([O-])=O

Specifications

Physical Form Liquid
Packaging Glass Bottle
Quantity 5 g
Color Colorless to Yellow
Deuterium (D) 98.5% min.
Linear Formula C6D5NO2
Formula Weight 128.15
Grade NMR
Chemical Name or Material Nitrobenzene-d5, 99 atom % D
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
Suspected of damaging fertility.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-014-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.