Learn More
Medchemexpress LLC HY-W010936 50mg Medchemexpress, Nitrobenzylthioinosine CAS:38048-32-7 Purity:>98%
SDP
Catalog No. 502083303
Encompass_Preferred
Medchemexpress, HY-W010936 50mg Nitrobenzylthioinosine CAS:38048-32-7 Nitrobenzylthioinosine is an ENT1 transporter inhibitor that binds to ENT1 transporter with high affinity. Nitrobenzylthioinosine is a photoaffinity probe for adenosine uptake sites in brain. Nitrobenzylthioinosine can cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3303 Supplier Medchemexpress LLC Supplier No. HYW01093650MG
Medchemexpress, HY-W010936 50mg Nitrobenzylthioinosine CAS:38048-32-7 Nitrobenzylthioinosine is an ENT1 transporter inhibitor that binds to ENT1 transporter with high affinity. Nitrobenzylthioinosine is a photoaffinity probe for adenosine uptake sites in brain. Nitrobenzylthioinosine can cross the blood-brain barrier. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.