Learn More
Apexbio Technology LLC Nitrocaramiphen hydrochloride 98636-73-8 50mg
SDP
Catalog No. 5000089650
Encompass_Preferred
Nitrocaramiphen hydrochloride (CAS 98636-73-8) is a small molecule that acts as a selective antagonist of muscarinic acetylcholine M1 receptors exhibiting a Ki of 5 5 nM Studies indicate that it effectively inhibits the hyperpolarizing effects induced by muscarinic agonists in muscle fibers Due to its receptor specificity nitrocaramiphen hydrochloride is widely utilized in neurobiological research to investigate cholinergic signaling pathways and the physiological roles of M1 receptor modulation
Catalog No. 50-000-89650 Supplier Apexbio Technology LLC Supplier No. B631150
Nitrocaramiphen hydrochloride (CAS 98636-73-8) is a small molecule that acts as a selective antagonist of muscarinic acetylcholine M1 receptors exhibiting a Ki of 5 5 nM Studies indicate that it effectively inhibits the hyperpolarizing effects induced by muscarinic agonists in muscle fibers Due to its receptor specificity nitrocaramiphen hydrochloride is widely utilized in neurobiological research to investigate cholinergic signaling pathways and the physiological roles of M1 receptor modulation