Learn More
Medchemexpress LLC Nitrocefin | 41906-86-9 | 92.3% | 516.50 | C21H16N4O8S2 | 5 MG
SDP
Catalog No. 5000264011
Encompass_Preferred
Nitrocefin is a chromogenic cephalosporin derivative used as a β-lactamase substrate for detecting and assaying β-lactamase activity. Hydrolysis of its β-lactam ring produces a visible color change, enabling rapid qualitative or quantitative measurement of β-lactamase enzymes in biochemical and microbiological assays.
  • Chromogenic substrate that produces a visible color change upon β-lactam hydrolysis.
  • Suitable for rapid qualitative and quantitative β-lactamase assays.
  • Powder form with defined low-temperature storage for extended stability.
  • High purity to support reproducible assay results.
  • Soluble in DMSO for preparation of concentrated stock solutions.
Catalog No. 50-002-64011 Supplier Medchemexpress LLC Supplier No. HY1089135MG
Nitrocefin is a chromogenic cephalosporin derivative used as a β-lactamase substrate for detecting and assaying β-lactamase activity. Hydrolysis of its β-lactam ring produces a visible color change, enabling rapid qualitative or quantitative measurement of β-lactamase enzymes in biochemical and microbiological assays.
  • Chromogenic substrate that produces a visible color change upon β-lactam hydrolysis.
  • Suitable for rapid qualitative and quantitative β-lactamase assays.
  • Powder form with defined low-temperature storage for extended stability.
  • High purity to support reproducible assay results.
  • Soluble in DMSO for preparation of concentrated stock solutions.