Learn More
Medchemexpress LLC Nitrocefin | 41906-86-9 | 98.9% | 516.50 | 1 ML
SDP
Catalog No. 5000345143
Encompass_Preferred
Nitrocefin is a highly activated, chromogenic cephalosporin derivative. It functions as a chromogenic β-lactamase substrate. Nitrocefin undergoes a distinct color change from yellow to red when the amide bond in its β-lactam ring is hydrolyzed by β-lactamase. It is used in competitive inhibition studies during the development of β-lactamase-resistant antibiotics.
  • Highly activated, chromogenic cephalosporin derivative
  • Chromogenic β-lactamase substrate
  • Undergoes a distinctive color change from yellow to red upon hydrolysis by β-lactamase
  • Used in competitive inhibition studies for β-lactamase-resistant antibiotics
  • Available in solid and solution forms
Catalog No. 50-003-45143 Supplier Medchemexpress LLC Supplier No. HY108913100MG
Nitrocefin is a highly activated, chromogenic cephalosporin derivative. It functions as a chromogenic β-lactamase substrate. Nitrocefin undergoes a distinct color change from yellow to red when the amide bond in its β-lactam ring is hydrolyzed by β-lactamase. It is used in competitive inhibition studies during the development of β-lactamase-resistant antibiotics.
  • Highly activated, chromogenic cephalosporin derivative
  • Chromogenic β-lactamase substrate
  • Undergoes a distinctive color change from yellow to red upon hydrolysis by β-lactamase
  • Used in competitive inhibition studies for β-lactamase-resistant antibiotics
  • Available in solid and solution forms