Learn More
Medchemexpress LLC Nitrocefin | 41906-86-9 | 98.9% | 516.50 | 50 MG
SDP
Catalog No. 5000346586
Encompass_Preferred
Nitrocefin is a highly activated, chromogenic cephalosporin derivative and a chromogenic β-lactamase substrate. It undergoes a distinctive color change from yellow to red as the amide bond in its β-lactam ring is hydrolyzed by β-lactamase. It is used in competitive inhibition studies in developmental work on β-lactamase-resistant antibiotics. It is intended for research use only.
  • Highly activated chromogenic cephalosporin derivative
  • Chromogenic β-lactamase substrate
  • Undergoes distinctive color change from yellow to red upon hydrolysis
  • Used in competitive inhibition studies for β-lactamase-resistant antibiotics
Catalog No. 50-003-46586 Supplier Medchemexpress LLC Supplier No. HY10891310MG
Nitrocefin is a highly activated, chromogenic cephalosporin derivative and a chromogenic β-lactamase substrate. It undergoes a distinctive color change from yellow to red as the amide bond in its β-lactam ring is hydrolyzed by β-lactamase. It is used in competitive inhibition studies in developmental work on β-lactamase-resistant antibiotics. It is intended for research use only.
  • Highly activated chromogenic cephalosporin derivative
  • Chromogenic β-lactamase substrate
  • Undergoes distinctive color change from yellow to red upon hydrolysis
  • Used in competitive inhibition studies for β-lactamase-resistant antibiotics