Learn More
TARGETMOL CHEMICALS INC NITROCEFIN 5MG
SDP
Catalog No. 502581045
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nitrocefin is an antibiotic. It being sensitive to hydrolysis by all lactamases produced by gram-positive and gram-negative bacteria. purity: 98%
Catalog No. 50-258-1045 Supplier TARGETMOL CHEMICALS INC Supplier No. T197095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nitrocefin is an antibiotic. It being sensitive to hydrolysis by all lactamases produced by gram-positive and gram-negative bacteria. purity: 98%