Learn More
Abcam Nitrocefin, Chromogenic cephalosporin beta-lactamase substrate, 5MG
Catalog No. 502215322
Encompass_Preferred
MW 516.5 Da, Purity >95%. Chromogenic cephalosporin β-lactamase substrate. Indicates β-lactamase activity via a red color intensely proportional to the original concentration of Nitrocefin. Undergoes rapid distinctive color change from yellow (max at pH 7.0 = 390 nm) to red (max at pH 7.0 = 486 nm) as the amide bond in the beta-lactam ring is hydrolyzed.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5322 Supplier Abcam Supplier No. AB1456255MG
MW 516.5 Da, Purity >95%. Chromogenic cephalosporin β-lactamase substrate. Indicates β-lactamase activity via a red color intensely proportional to the original concentration of Nitrocefin. Undergoes rapid distinctive color change from yellow (max at pH 7.0 = 390 nm) to red (max at pH 7.0 = 486 nm) as the amide bond in the beta-lactam ring is hydrolyzed.
The product is subject to the following: Abcam Restricted Use Statement