Learn More
Abcam Nitrocellulose Transfer Membrane- 0.45 µm, 10MEMBRANE
Supplier:  Abcam AB13341210MEMBRANE
Encompass_Preferred
High quality 0.45 um nitrocellulose membranes that are more durable than standard nitrocellulose membranes. ab133412 yields clearer blots as the membrane is less prone to distortion.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7017