Learn More
Parker Hannifan Parker Hannifin NitroFlow 60 Generator, High-Purity Nitrogen, Oil-less Compressor, 60 SLPM
SDP
Supplier:  Parker Hannifan NITROFLOW60NA
Encompass
The PARKER HANNIFIN NITROFLOW 60NA is a nitrogen gas generator designed for laboratory applications, providing high-purity nitrogen on-demand to enhance safety and efficiency. This self-contained system eliminates the need for gas cylinders and operates quietly. It features an oil-less rotary scroll compressor and multi-stage filtration to deliver clean, dry gas, making it suitable for various analytical tasks such as nebulization and instrument purging.
Catalog No. NC3760812
Only null left
Add to Cart
Request a Quote