Learn More
Alkali Scientific Nitrofurantoin, 1 G
SDP
Catalog No. 5000661514
Encompass_Preferred
Nitrofurantoin, 1g, is a laboratory-grade antibiotic used in the treatment and study of urinary tract infections (UTIs) caused by susceptible bacteria. As a broad-spectrum antimicrobial agent, it is effective against both Gram-positive and Gram-negative bacteria. Nitrofurantoin works by disrupting bacterial protein synthesis, DNA synthesis, and metabolism, which leads to bacterial death. This 1g quantity is ideal for use in laboratory tests, including susceptibility testing and other microbial assays. Nitrofurantoin is frequently used in clinical microbiology to assess bacterial resistance patterns and study antimicrobial resistance mechanisms.
Catalog No. 50-006-61514 Supplier Alkali Scientific Supplier No. NIT001CP
Nitrofurantoin, 1g, is a laboratory-grade antibiotic used in the treatment and study of urinary tract infections (UTIs) caused by susceptible bacteria. As a broad-spectrum antimicrobial agent, it is effective against both Gram-positive and Gram-negative bacteria. Nitrofurantoin works by disrupting bacterial protein synthesis, DNA synthesis, and metabolism, which leads to bacterial death. This 1g quantity is ideal for use in laboratory tests, including susceptibility testing and other microbial assays. Nitrofurantoin is frequently used in clinical microbiology to assess bacterial resistance patterns and study antimicrobial resistance mechanisms.