Learn More
Medchemexpress LLC Nitrofurantoin-13c3 | 1217226-46-4 | C₅¹³C₃H₆N₄O₅ | 1 MG
SDP
Catalog No. 5000432055
Encompass_Preferred
Nitrofurantoin-13C3 is the 13C labeled Nitrofurantoin. Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes (carbon) for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-32055 Supplier Medchemexpress LLC Supplier No. HYA0090S1MG
Nitrofurantoin-13C3 is the 13C labeled Nitrofurantoin. Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes (carbon) for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.