Learn More
Alkali Scientific Nitrofurantoin, 25 G
SDP
Catalog No. 5000661518
Encompass_Preferred
Nitrofurantoin, 25g, is a broad-spectrum antibiotic used for the treatment of urinary tract infections (UTIs) and for laboratory research in microbiology. It is effective against a variety of bacteria, including both Gram-positive and Gram-negative organisms, by inhibiting bacterial DNA and protein synthesis. This 25g quantity is suitable for larger-scale antimicrobial studies and for laboratories requiring significant amounts of the antibiotic for routine susceptibility testing. It is commonly used in clinical and microbiological laboratories for testing bacterial resistance, particularly for urinary pathogens and in developing resistance profiles for clinical diagnostics.
Catalog No. 50-006-61518 Supplier Alkali Scientific Supplier No. NIT002CP
Nitrofurantoin, 25g, is a broad-spectrum antibiotic used for the treatment of urinary tract infections (UTIs) and for laboratory research in microbiology. It is effective against a variety of bacteria, including both Gram-positive and Gram-negative organisms, by inhibiting bacterial DNA and protein synthesis. This 25g quantity is suitable for larger-scale antimicrobial studies and for laboratories requiring significant amounts of the antibiotic for routine susceptibility testing. It is commonly used in clinical and microbiological laboratories for testing bacterial resistance, particularly for urinary pathogens and in developing resistance profiles for clinical diagnostics.