Learn More
Selleck Chemical LLC Nitrofurantoin S4536-100mg
SDP
Catalog No. 503110312
Encompass_Preferred
Nitrofurantoin (Macrodantin Furadantine Furadonine) is an antibiotic used to treat bladder infections It inhibits bacterial DNA RNA and cell wall protein synthesis
Catalog No. 50-311-0312 Supplier Selleck Chemical LLC Supplier No. S4536100MG
Nitrofurantoin (Macrodantin Furadantine Furadonine) is an antibiotic used to treat bladder infections It inhibits bacterial DNA RNA and cell wall protein synthesis