Learn More
Alkali Scientific Nitrofurantoin, 5 G
SDP
Catalog No. 5000661345
Encompass_Preferred
Nitrofurantoin, 5g, is a laboratory-grade antibiotic used to study bacterial resistance, particularly in the context of urinary tract infections (UTIs). It is effective against a broad spectrum of Gram-positive and Gram-negative bacteria by disrupting their protein synthesis, DNA replication, and overall metabolism. The 5g quantity is ideal for use in antimicrobial susceptibility testing, such as Minimum Inhibitory Concentration (MIC) assays, disk diffusion, and other microbiological tests. Nitrofurantoin is also used in research settings to better understand antimicrobial resistance mechanisms, including the development of resistance in UTI pathogens.
Catalog No. 50-006-61345 Supplier Alkali Scientific Supplier No. NIT003CP
Nitrofurantoin, 5g, is a laboratory-grade antibiotic used to study bacterial resistance, particularly in the context of urinary tract infections (UTIs). It is effective against a broad spectrum of Gram-positive and Gram-negative bacteria by disrupting their protein synthesis, DNA replication, and overall metabolism. The 5g quantity is ideal for use in antimicrobial susceptibility testing, such as Minimum Inhibitory Concentration (MIC) assays, disk diffusion, and other microbiological tests. Nitrofurantoin is also used in research settings to better understand antimicrobial resistance mechanisms, including the development of resistance in UTI pathogens.