Learn More
Research Products International Corp Nitrofurantoin, 5 G
SDP
Catalog No. 501978373
Encompass_Preferred
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis. 
Researchers may use nitrofurantoin as a selective agent by incorporating it into culture media to promote the growth of cells or organisms that have been genetically engineered to express resistance to this antibiotic. It may have applications in microbiology and cell culture experiments, particularly in the context of antibiotic selection.
WARNING! This product can expose you to Nitrofurantoin, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-8373 Supplier Research Products International Corp Supplier No. N515005.0
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis. 
Researchers may use nitrofurantoin as a selective agent by incorporating it into culture media to promote the growth of cells or organisms that have been genetically engineered to express resistance to this antibiotic. It may have applications in microbiology and cell culture experiments, particularly in the context of antibiotic selection.
WARNING! This product can expose you to Nitrofurantoin, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.