Learn More
Medchemexpress LLC Nitrofurantoin | 67-20-9 | 238.16 | 5 G
SDP
Catalog No. 5000365968
Encompass_Preferred
Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Potent and orally active broad-spectrum beta-lactamase antimicrobial agent
  • Acts as an antibiotic
  • Used for the study of urinary tract infections (UTIs), including cystitis and kidney infections
  • Targets bacteria
  • Melting/freezing point of 268°C
Catalog No. 50-003-65968 Supplier Medchemexpress LLC Supplier No. 5GHYA0090
Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Potent and orally active broad-spectrum beta-lactamase antimicrobial agent
  • Acts as an antibiotic
  • Used for the study of urinary tract infections (UTIs), including cystitis and kidney infections
  • Targets bacteria
  • Melting/freezing point of 268°C