Learn More
Medchemexpress LLC Nitrofurantoin | 67-20-9 | C8H6N4O5 | 1 G
SDP
Catalog No. 5000432779
Encompass_Preferred
Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Potent and orally active
  • Broad-spectrum beta-lactamase antimicrobial agent
  • Acts as an antibiotic
  • Used for the study of urinary tract infections (UTIs), including cystitis and kidney infections
Catalog No. 50-004-32779 Supplier Medchemexpress LLC Supplier No. HYA00901G
Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Potent and orally active
  • Broad-spectrum beta-lactamase antimicrobial agent
  • Acts as an antibiotic
  • Used for the study of urinary tract infections (UTIs), including cystitis and kidney infections