Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitrofurantoin | 67-20-9 | MFCD00003224 | 99.7% | C8H6N4O5 | 1 ML
SDP

Catalog No. 5000432176
Encompass_Preferred

Nitrofurantoin is a potent, orally active, broad-spectrum beta-lactamase antimicrobial agent. It functions as an antibiotic, making it suitable for research into urinary tract infections (UTIs), including cystitis and kidney infections.

  • Potent broad-spectrum antimicrobial agent
  • Antibiotic for urinary tract infection research
  • Purity of 99.7%
  • Soluble in DMSO (100 mg/mL)
  • Light yellow to yellow solid appearance
  • For research use only

Catalog No. 50-004-32176 Supplier Medchemexpress LLC Supplier No. HYA009010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrofurantoin is a potent, orally active, broad-spectrum beta-lactamase antimicrobial agent. It functions as an antibiotic, making it suitable for research into urinary tract infections (UTIs), including cystitis and kidney infections.

  • Potent broad-spectrum antimicrobial agent
  • Antibiotic for urinary tract infection research
  • Purity of 99.7%
  • Soluble in DMSO (100 mg/mL)
  • Light yellow to yellow solid appearance
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.