Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrofurantoin, 99.5%, MP Biomedicals™

Catalog No. ICN15588180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15588180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15588180 Supplier MP Biomedicals, Inc Supplier No. 0215588180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-20-9
Molecular Formula C8H6N4O5
Molecular Weight (g/mol) 238.16
MDL Number MFCD00003224
InChI Key NXFQHRVNIOXGAQ-YCRREMRBSA-N
Synonym nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
PubChem CID 6604200
ChEBI CHEBI:71415
IUPAC Name 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O

Specifications

Color Orange-Yellow to Yellow
Assay Percent Range 0.995
Quantity 100 g
Formula Weight 238.2
Physical Form Crystalline Powder
Chemical Name or Material Nitrofurantoin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.