Learn More
Medchemexpress LLC Nitrofurantoin (standard) | 67-20-9 | 99.7% | C8H6N4O5 | 100 MG
SDP
Catalog No. 5000432184
Encompass_Preferred
Nitrofurantoin (Standard) is the analytical standard for Nitrofurantoin, intended for research and analytical applications. It is a potent, orally active, broad-spectrum beta-lactamase antimicrobial agent. This antibiotic is used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Analytical standard for research and analytical applications
  • Potent and orally active antimicrobial agent
  • Broad-spectrum beta-lactamase activity
  • Functions as an antibiotic
  • Used for studying urinary tract infections (UTIs), cystitis, and kidney infections
  • Employed in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS
Catalog No. 50-004-32184 Supplier Medchemexpress LLC Supplier No. HYA0090R100MG
Nitrofurantoin (Standard) is the analytical standard for Nitrofurantoin, intended for research and analytical applications. It is a potent, orally active, broad-spectrum beta-lactamase antimicrobial agent. This antibiotic is used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Analytical standard for research and analytical applications
  • Potent and orally active antimicrobial agent
  • Broad-spectrum beta-lactamase activity
  • Functions as an antibiotic
  • Used for studying urinary tract infections (UTIs), cystitis, and kidney infections
  • Employed in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS