Learn More
Medchemexpress LLC Nitrofurantoin (Standard) | 67-20-9 | 99.7% | C8H6N4O5 | 250 MG
SDP
Catalog No. 5000358585
Encompass_Preferred
Nitrofurantoin (Standard) is the analytical standard of Nitrofurantoin, intended for research and analytical applications. It is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Analytical standard grade, supplied as an assay reference
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-003-58585 Supplier Medchemexpress LLC Supplier No. 250MGHYA0090R
Nitrofurantoin (Standard) is the analytical standard of Nitrofurantoin, intended for research and analytical applications. It is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. It acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Analytical standard grade, supplied as an assay reference
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS