Learn More
Medchemexpress LLC Nitrofurantoin (Standard) | 67-20-9 | 99.7% | C8H6N4O5 | 50 MG
SDP
Catalog No. 5000433559
Encompass_Preferred
Nitrofurantoin (Standard) is the analytical standard of Nitrofurantoin. It is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent, primarily used for research and analytical applications. This antibiotic is suitable for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Analytical standard for qualitative, quantitative, and methodological research
  • Used in HPLC, GC, and MS experiments
  • Potent and orally active broad-spectrum antimicrobial agent
  • Acts as an antibiotic
  • Suitable for studying urinary tract infections, including cystitis and kidney infections
Catalog No. 50-004-33559 Supplier Medchemexpress LLC Supplier No. HYA0090R50MG
Nitrofurantoin (Standard) is the analytical standard of Nitrofurantoin. It is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent, primarily used for research and analytical applications. This antibiotic is suitable for the study of urinary tract infections (UTIs), including cystitis and kidney infections.
  • Analytical standard for qualitative, quantitative, and methodological research
  • Used in HPLC, GC, and MS experiments
  • Potent and orally active broad-spectrum antimicrobial agent
  • Acts as an antibiotic
  • Suitable for studying urinary tract infections, including cystitis and kidney infections