Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrofurantoin, USP Grade, G-Biosciences
SCI_ED

Catalog No. S157217
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157217 250 g
S139786 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157217 Supplier G-Biosciences Supplier No. RC1069
Add to Cart
Add to Cart

Nitrofurantoin is a bacteriostatic, nitrofuran antibiotic targeting gram-negative and gram-positive bacteria such as Enterococci, Staphylococci, Streptococci, Corynebacterium, and E. Coli.

It has been used to study nitrofurantoin bacterial susceptibility and nitrofurantoin-induced toxicity. Additionally, it is a substrate for bacterial nitrofuran reductase.

Chemical Identifiers

Molecular Weight (g/mol) 238.16 g/mol

Specifications

Boiling Point 380.75°C
Quantity 250 g
Percent Purity >98%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.