Learn More
Medchemexpress LLC Nitrofurazone-13C,15N2 | 1217220-85-3 | 99.6% | 1 MG
SDP
Catalog No. 5000405441
Encompass_Preferred
Nitrofurazone-13C,15N2 is the 13C and 15N labeled Nitrofurazone, a potential antibiotic used topically for wounds, burns, ulcers, and skin infections. It combats various microorganisms, assists in surfactant preparation, and may impact marine ecosystem function and epiphytic fauna. This product is for research use only. Stable heavy isotopes like carbon and nitrogen are incorporated into drug molecules as tracers for quantitation during development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Can be applied topically for treating wounds, burns, ulcers, and skin infections
  • Used to combat various microorganisms and in the preparation of surfactants
  • May influence the ecosystem function in marine environments and the functional processes of epiphytic fauna
Catalog No. 50-004-05441 Supplier Medchemexpress LLC Supplier No. HYB0226S1MG
Nitrofurazone-13C,15N2 is the 13C and 15N labeled Nitrofurazone, a potential antibiotic used topically for wounds, burns, ulcers, and skin infections. It combats various microorganisms, assists in surfactant preparation, and may impact marine ecosystem function and epiphytic fauna. This product is for research use only. Stable heavy isotopes like carbon and nitrogen are incorporated into drug molecules as tracers for quantitation during development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Can be applied topically for treating wounds, burns, ulcers, and skin infections
  • Used to combat various microorganisms and in the preparation of surfactants
  • May influence the ecosystem function in marine environments and the functional processes of epiphytic fauna