Learn More
Medchemexpress LLC Nitrofurazone-13C,15N2 | 1217220-85-3 | 99.6% | 5 MG
SDP
Catalog No. 5000405475
Encompass_Preferred
Nitrofurazone-13C,15N2, also known as Nitrofural-13C,15N2, is a 13C and 15N labeled derivative of Nitrofurazone. This antibiotic is used topically to treat wounds, burns, ulcers, and skin infections, and in surfactant preparation. It may also affect ecosystem function in marine environments. The stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Supports studies on pharmacokinetic and metabolic profiles of drugs
  • Aids in combating various microorganisms
  • Applied in the preparation of surfactants
Catalog No. 50-004-05475 Supplier Medchemexpress LLC Supplier No. HYB0226S5MG
Nitrofurazone-13C,15N2, also known as Nitrofural-13C,15N2, is a 13C and 15N labeled derivative of Nitrofurazone. This antibiotic is used topically to treat wounds, burns, ulcers, and skin infections, and in surfactant preparation. It may also affect ecosystem function in marine environments. The stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Supports studies on pharmacokinetic and metabolic profiles of drugs
  • Aids in combating various microorganisms
  • Applied in the preparation of surfactants