Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitrofurazone | 59-87-0 | 99.96% | 1 ML
SDP

Catalog No. 5000406130
Encompass_Preferred

Nitrofurazone (Nitrofural) is a broad-spectrum, nitro-aromatic antibiotic with oral activity. It is effective against both Gram-positive and Gram-negative bacteria, making it suitable for research into various infections and skin conditions.

  • Active against Gram-positive and Gram-negative bacteria.
  • Useful in research for various infections and skin conditions.
  • Inhibits RNA and ribosomal subunit synthesis.
  • Can be used to create breast tumor models.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Storage in solvent: -80°C for 2 years; -20°C for 1 year.

Catalog No. 50-004-06130 Supplier Medchemexpress LLC Supplier No. HYB022610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrofurazone (Nitrofural) is a broad-spectrum, nitro-aromatic antibiotic with oral activity. It is effective against both Gram-positive and Gram-negative bacteria, making it suitable for research into various infections and skin conditions.

  • Active against Gram-positive and Gram-negative bacteria.
  • Useful in research for various infections and skin conditions.
  • Inhibits RNA and ribosomal subunit synthesis.
  • Can be used to create breast tumor models.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Storage in solvent: -80°C for 2 years; -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.