Learn More
Medchemexpress LLC Nitrofurazone (Nitrofural) | 59-87-0 | 99.96% | 5 G
SDP
Catalog No. 5000406129
Encompass_Preferred
Nitrofurazone (Nitrofural) is a broad-spectrum antibiotic with oral activity. It is a nitro-aromatic drug active against both Gram-positive and Gram-negative bacteria. This product is for research use only.
  • Broad-spectrum antibiotic
  • Oral activity
  • Nitro-aromatic drug
  • Active against both Gram-positive and Gram-negative bacteria
Catalog No. 50-004-06129 Supplier Medchemexpress LLC Supplier No. HYB02265G
Nitrofurazone (Nitrofural) is a broad-spectrum antibiotic with oral activity. It is a nitro-aromatic drug active against both Gram-positive and Gram-negative bacteria. This product is for research use only.
  • Broad-spectrum antibiotic
  • Oral activity
  • Nitro-aromatic drug
  • Active against both Gram-positive and Gram-negative bacteria