Learn More
Medchemexpress LLC Nitrofurazone (Nitrofural) | 59-87-0 | MFCD00003225 | 99.96% | 1 G
SDP
Catalog No. 5000407213
Encompass_Preferred
Nitrofurazone (Nitrofural) is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria. It is a nitrofuran antibiotic primarily used topically for various bacterial skin infections, burns, ulcers, and wounds. It retains its antibacterial activity in blood, serum, and pus, and does not interfere with healing.
  • Broad spectrum antibiotic
  • Active against Gram-positive and Gram-negative bacteria
  • Used for bacterial skin infections, burns, ulcers, and wounds
  • Maintains antibacterial activity in biological fluids
  • Does not inhibit phagocytosis
  • Does not interfere with healing
Catalog No. 50-004-07213 Supplier Medchemexpress LLC Supplier No. HYB02261G
Nitrofurazone (Nitrofural) is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria. It is a nitrofuran antibiotic primarily used topically for various bacterial skin infections, burns, ulcers, and wounds. It retains its antibacterial activity in blood, serum, and pus, and does not interfere with healing.
  • Broad spectrum antibiotic
  • Active against Gram-positive and Gram-negative bacteria
  • Used for bacterial skin infections, burns, ulcers, and wounds
  • Maintains antibacterial activity in biological fluids
  • Does not inhibit phagocytosis
  • Does not interfere with healing