Learn More
Chemglass Life Sciences Nitrogen Flowmeter, 0.04 to 0.5L/min
SDP
Catalog No. NC1550730
Encompass_Preferred
Precision machined clear acrylic Nitrogen flowmeter with Brass metering valve is for use with Chemglass process reactors. Front of flowmeter has a blue scale indicating the L/min of air. HDPE hose connection is for use with 5/16” ID tubing. Unit is supplied complete with a 1/2” OD mounting arm for ease in clamping to support frame.
Catalog No. NC1550730 Supplier Chemglass Life Sciences Supplier No. CG1605A10
Precision machined clear acrylic Nitrogen flowmeter with Brass metering valve is for use with Chemglass process reactors. Front of flowmeter has a blue scale indicating the L/min of air. HDPE hose connection is for use with 5/16” ID tubing. Unit is supplied complete with a 1/2” OD mounting arm for ease in clamping to support frame.