Learn More
Organomation NITRO-GEN+ Nitrogen Generator
SDP
Supplier:  Organomation NA1935
Encompass_Preferred
The NITRO-GEN+ Nitrogen Generator (Catalog# NA1935) is compatible with all Organomation nitrogen evaporators up to 100 sample positions, and is designed to fulfill the needs of sample preparation and LC-MS analysis. It is a self-contained generator that is able to produce up to 35 LPM of nitrogen gas with a purity up to 97%. This generator produces nitrogen by combining dual compressor and Carbon Molecular Sieve (CMS) technology. Its dual compressor technology utilizes both a high and low pressure compressor, both of which are included with the unit. This allows the unit to remain quiet while also reducing the stress on the compressors, extending their life and reducing maintenance costs. This generator is able to produce nitrogen at both a low pressure and ambient temperature, eliminating the need for hazardous high pressure cylinders. The NITRO-GEN+ promotes a safe laboratory environment while being a reliable source of N2 gas for a variety of applications.
Catalog No. NC2330630
Add to Cart
Request a Quote