Learn More
Andon Specialties NITROGEN GUN
SDP
Catalog No. NC0463894
Encompass
Nitrogen Spray Gun, Natural Nozzle Color, 3/16 in. orifice, 1/4 in. FNPT Connection, All wetted parts PFA, PTFE, and Polyethylene
Catalog No. NC0463894 Supplier Andon Specialties Supplier No. 4214211
Nitrogen Spray Gun, Natural Nozzle Color, 3/16 in. orifice, 1/4 in. FNPT Connection, All wetted parts PFA, PTFE, and Polyethylene