Learn More
Hach Company Nitrate Standard Solution, 1 mg/L, 500 mL
Supplier:  Hach Company 204649
Encompass
ISE Standard Solution. Nitrate, 1.00 mg/L ±0.01 mg/L as NO3-N (NIST). 500 mL bottle
Catalog No. 50-199-4669