Learn More
Electron Microscopy Sciences Nitrogen Purge Cabinet Purge Cabinet Regular, 16x9, 10.5 in height
SDP
Catalog No. 50930906
Encompass_Preferred
Nitrogen purge cabinets can be used at at temperatures up to 120 degree F in freezers, refrigerators, or at room temperature. Cabinets have airtight doors that need no lubrication, sliding perforated shelves for easy access, and transparent acrylic wall for see-through storage. Acrylic, ethylene propylene, and Type 316 stainless steel welded parts.
Footprint: 16 in x 9 in
Catalog No. 50-930-906 Supplier Electron Microscopy Sciences Supplier No. 7121920
Nitrogen purge cabinets can be used at at temperatures up to 120 degree F in freezers, refrigerators, or at room temperature. Cabinets have airtight doors that need no lubrication, sliding perforated shelves for easy access, and transparent acrylic wall for see-through storage. Acrylic, ethylene propylene, and Type 316 stainless steel welded parts.
Footprint: 16 in x 9 in