Learn More
Electron Microscopy Sciences Nitrogen Purge Cabinet Purge Cabinet with Flow Meter, 8.5 in height
SDP
Catalog No. 50930907
Encompass_Preferred
Nitrogen purge cabinets can be used at at temperatures up to 120 degree F in freezers, refrigerators, or at room temperature. Cabinets have airtight doors that need no lubrication, sliding perforated shelves for easy access, and transparent acrylic wall for see-through storage. Acrylic, ethylene propylene, and Type 316 stainless steel welded parts. Direct reading scale flow meter with control levels allow flow adjustments in the line.
Footprint: 10 in x 7 in
Catalog No. 50-930-907 Supplier Electron Microscopy Sciences Supplier No. 7121930
Nitrogen purge cabinets can be used at at temperatures up to 120 degree F in freezers, refrigerators, or at room temperature. Cabinets have airtight doors that need no lubrication, sliding perforated shelves for easy access, and transparent acrylic wall for see-through storage. Acrylic, ethylene propylene, and Type 316 stainless steel welded parts. Direct reading scale flow meter with control levels allow flow adjustments in the line.
Footprint: 10 in x 7 in