Learn More
Aqua Solutions Nitrogen Standard 100 ppm | 7727-37-9 | MFCD00011416 | 500 mL
SDP
Supplier:  Aqua Solutions 6494500ML
Encompass_Preferred
Nitrogen Standard 100 ppm | Mol Wt: 28.01 | 7727-37-9 | MFCD00011416 | 500 mL. Standard good for 60 days.
Catalog No. NC1029962
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov