Learn More
SPEX CERTIPREP LLC 3P NITROGEN STANDARD 100PPM 500ML
Catalog No. NC9869983
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Nitrogen from NaNO2 in water, 99.99+% (metals basis); Single Component IC Standard, 12 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9869983 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICNO2500
(HPS by Zeptometrix) 100 ug/mL Nitrogen from NaNO2 in water, 99.99+% (metals basis); Single Component IC Standard, 12 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.