Learn More
Hach Company Nitrogen-Nitrate Standard Solution, 15 mg/L as NO3-N (NIST), 100 mL MDB
Catalog No. 501995950 Shop All Hach Company Products
Encompass
For accuracy checks in nitrate nitrogen determinations
Catalog No. 50-199-5950 Supplier Hach Company Supplier No. 2415132
For accuracy checks in nitrate nitrogen determinations