Learn More
Strem, An Ascensus Company CAS# 175166-51-5. 100mg. (3aS,3'aS,8aR,8'aR)-2,2'-(1-Methylethylidene)bis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole], 98%, (99% ee).
SDP
Supplier:  Strem, An Ascensus Company 071034100MG
Encompass
CAS# 175166-51-5. 100mg. (3aS,3'aS,8aR,8'aR)-2,2'-(1-Methylethylidene)bis[3a,8a-dihydro-8H-indeno[1,2-d]oxazole], 98%, (99% ee). . Molecular Weight: 358.40. Molecular Formula: C23H22N2O2. Color/form: gray pwdr. Strem# 07-1034. http://www.strem.com/catalog/v/07-1034/
Catalog No. NC1479318