Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitroguanidine, Moistened with ca 25% Water 99%, Thermo Scientific™

Catalog No. AC128671000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC128671000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC128671000 Supplier Thermo Scientific Chemicals Supplier No. 128671000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-88-7 , 7732-18-5
Molecular Formula CH4N4O2
Molecular Weight (g/mol) 104.07
MDL Number MFCD00007039
InChI Key IDCPFAYURAQKDZ-UHFFFAOYSA-N
Synonym nitroguanidine, 1-nitroguanidine, n-nitroguanidine, picrite, guanidine, nitro, guanidine, 1-nitro, picrite the explosive, guanidine, n-nitro, guanidine, 1-nitro-wet
PubChem CID 11174
ChEBI CHEBI:39180
IUPAC Name N''-nitroguanidine
SMILES NC(N)=N[N+]([O-])=O

Specifications

Acidity 0.06% max. (H2SO4)
CAS Min % 98.5
CAS Max % 100.0
Color White
pH 4.5 to 7
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula O2NNHC(=NH)NH2
Quantity 100 g
Beilstein 03, 126
Merck Index 15, 6695
Solubility Information 0.2% max. Insoluble in Water
Formula Weight 104.07
Percent Purity ≥98.5% (on dry substance)
Physical Form Powder
Chemical Name or Material Nitroguanidine, 99%
Show More Show Less
Hazard Category FLAMMABLE SOLID
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.